accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClCH3HN(S)NH2NSHH

CAS RN 104632-25-9

IUPAC Name: dihydrogen (6S)-N6-propyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine dichloride
Synonyms: dihydrogen pramipexole dichloride
Molecular Weight (g/mol): 284.24
Molecular Formula: C10H19Cl2N3S
InChi Key: QMNWXHSYPXQFSK-KLXURFKVSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CCCN[C@H]1CCC2=C(C1)SC(N)=N2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)