Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Pramipexole dihydrochloride
GREENER_CHOICESDP

Catalog No. 417450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-745-0 50 mg
41-741-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-745-0 Supplier R&D Systems™ Supplier No. 4174/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective D3 agonist

Dopamine receptor agonist exhibiting selectivity for the D3 receptor (Ki values are 0.5, 3.3, 3.9 and 3.9 nM for D3, D2S, D2L and D4 receptors respectively). Exhibits negligible affinity for D1 and D5 receptors. Displays activity in the treatment of Parkinson's disease.

Chemical Identifiers

CAS 104632-25-9
Molecular Formula C10H19Cl2N3S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 284.25
Purity >99%
Molecular Weight (g/mol) 284.243
Chemical Name or Material Pramipexole dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.