CAS RN 104869-26-3
IUPAC Name:
hexasodium {[(8-{4-fluoro-3-[3-({[3-({2-fluoro-5-[(4,6,8-trisulfonatonaphthalen-1-yl)carbamoyl]phenyl}carbamoyl)phenyl]carbamoyl}amino)benzamido]benzamido}-3,5-disulfonatonaphthalen-1-yl)sulfanyl]peroxy}olate
Synonyms:
hexasodium [(8-{4-fluoro-3-[3-({[3-({2-fluoro-5-[(4,6,8-trisulfonatonaphthalen-1-yl)carbamoyl]phenyl}carbamoyl)phenyl]carbamoyl}amino)benzamido]benzamido}-3,5-disulfonatonaphthalen-1-yl)sulfanyl]peroxyolate
Molecular Weight (g/mol):
1437.08
Molecular Formula:
C49H28F2N6Na6O23S6
InChi Key:
QJFCMJZVZFAWGF-UHFFFAOYSA-H
SMILES:
[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[O-]OOSC1=C2C(NC(=O)C3=CC(NC(=O)C4=CC(NC(=O)NC5=CC=CC(=C5)C(=O)NC5=C(F)C=CC(=C5)C(=O)NC5=CC=C(C6=CC(=CC(=C56)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O)=CC=C4)=C(F)C=C3)=CC=C(C2=CC(=C1)S([O-])(=O)=O)S([O-])(=O)=O
1
–
1
of
1
results
1
Tocris Bioscience™ NF 157
CAS: 104869-26-3 Molecular Formula: C49H28F2N6Na6O23S6