accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNClONOSN

CAS RN 105219-56-5

IUPAC Name: 3-[7-(2-chlorophenyl)-13-methyl-3-thia-1,8,11,12-tetraazatricyclo[8.3.0.0²,⁶]trideca-2(6),4,7,10,12-pentaen-4-yl]-1-(morpholin-4-yl)propan-1-one
Synonyms: 3-[7-(2-chlorophenyl)-13-methyl-3-thia-1,8,11,12-tetraazatricyclo[8.3.0.0²,⁶]trideca-2(6),4,7,10,12-pentaen-4-yl]-1-(morpholin-4-yl)propan-1-one
Molecular Weight (g/mol): 455.96
Molecular Formula: C22H22ClN5O2S
InChi Key: JGPJQFOROWSRRS-UHFFFAOYSA-N
SMILES: CC1=NN=C2CN=C(C3=C(SC(CCC(=O)N4CCOCC4)=C3)N12)C1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)