Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ WEB 2086
GREENER_CHOICESDP

Catalog No. 233910 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-391-0 10 mg
23-391 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-391-0 Supplier R&D Systems™ Supplier No. 2339/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent PAF receptor antagonist

Potent, selective platelet-activating factor (PAF) receptor antagonist (Ki = 16.3 nM). Displays anti-inflammatory, antiangiogenic and anticancer activity. Inhibits growth and proliferation of MCF-7 breast cancer cells.

Chemical Identifiers

CAS 105219-56-5
Molecular Formula C22H22ClN5O2S

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 455.96
Purity >99%
Molecular Weight (g/mol) 455.961
Chemical Name or Material WEB 2086
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.