Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Tamoxifen
SDP

Catalog No. RD10195121 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10195121 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10195121 Supplier R&D Systems™ Supplier No. 6342/100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Tocris product, part of R&D Systems. Estrogen receptor partial agonist/antagonist

Tamoxifen is a selective estrogen receptor modulator (SERM). Exhibits antiestrogenic effects in mammary tissue and estrogen-like effects in bone. Anticancer. Tamoxifen citrate (Cat.No. 0999) is also available.

Specifications

Chemical Name or Material 2-[4-[(1Z)-1,2-Diphenyl-1-buten-1-yl]phenoxy]-N,N-dimethylethanamine
CAS 10540-29-1
Quantity 100 mg
Product Line Tocris
Target Estrogen and Related Receptor Modulators
Molecular Formula C26H29NO
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.