CAS RN 105637-50-1
IUPAC Name:
hydrogen 1-[(5-chloronaphthalen-1-yl)sulfonyl]-1,4-diazepane chloride
Synonyms:
hydrogen 1-(5-chloronaphthalen-1-ylsulfonyl)-1,4-diazepane chloride
Molecular Weight (g/mol):
361.28
Molecular Formula:
C15H18Cl2N2O2S
InChi Key:
ZNRYCIVTNLZOGI-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].ClC1=C2C=CC=C(C2=CC=C1)S(=O)(=O)N1CCCNCC1
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ ML-9, Hydrochloride
A cell-permeable, reversible and ATP-competitive inhibitor of myosin light chain kinase, proteins kinase A, and protein kinase C
2
R&D Systems™ ML 9 hydrochloride
CAS: 105637-50-1 Molecular Formula: C15H18Cl2N2O2S