Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ML 9 hydrochloride
GREENER_CHOICESDP

Catalog No. 043110 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-311-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 04-311-0 Supplier R&D Systems™ Supplier No. 0431/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Myosin light chain kinase inhibitor

ML 9 hydrochloride is a selective MLCK inhibitor (Ki values are 4, 32 and 54 μM for MLCK, PKA and PKC respectively). Also inhibits STIM1-plasma membrane interactions, preventing store-operated Ca2+ entry (SOCE).

Chemical Identifiers

CAS 105637-50-1
Molecular Formula C15H18Cl2N2O2S

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 361.29
Purity >99%
Molecular Weight (g/mol) 361.281
Chemical Name or Material ML 9 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.