CAS RN 106266-06-2
Risperidone, MP Biomedicals
CAS: 106266-06-2 Molecular Formula: C23H27FN4O2 Molecular Weight (g/mol): 410.493 InChI Key: RAPZEAPATHNIPO-UHFFFAOYSA-N Synonym: risperidone,risperidal,risperdal,risperdal consta,rispolept,risperin,rispolin,sequinan,risperidona,risperidonum PubChem CID: 5073 ChEBI: CHEBI:8871 IUPAC Name: 3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one SMILES: CC1=C(C(=O)N2CCCCC2=N1)CCN3CCC(CC3)C4=NOC5=C4C=CC(=C5)F
Risperidone Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Risperidone is a new central 5-hydroxytryptamine2 and dopamine D2 antagonist. It is used to treat agitation, aggression,and psychosis caused during demnetia. It is a neuroleptic drug. It is also an atypical antipsychotic drug which is used for treating schizophrenia.
Risperidone, 99%
CAS: 106266-06-2 Molecular Formula: C23H27FN4O2 Molecular Weight (g/mol): 410.49 InChI Key: RAPZEAPATHNIPO-UHFFFAOYSA-N Synonym: risperidone,risperidal,risperdal,risperdal consta,rispolept,risperin,rispolin,sequinan,risperidona,risperidonum PubChem CID: 5073 ChEBI: CHEBI:8871 IUPAC Name: 3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one SMILES: CC1=C(C(=O)N2CCCCC2=N1)CCN3CCC(CC3)C4=NOC5=C4C=CC(=C5)F
Risperidone, 100.7%, MP Biomedicals™
CAS: 106266-06-2 Molecular Formula: C23H27FN4O2 Molecular Weight (g/mol): 410.493 InChI Key: RAPZEAPATHNIPO-UHFFFAOYSA-N Synonym: risperidone,risperidal,risperdal,risperdal consta,rispolept,risperin,rispolin,sequinan,risperidona,risperidonum PubChem CID: 5073 ChEBI: CHEBI:8871 IUPAC Name: 3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one SMILES: CC1=C(C(=O)N2CCCCC2=N1)CCN3CCC(CC3)C4=NOC5=C4C=CC(=C5)F
Risperidone 98.0+%, TCI America™
CAS: 106266-06-2 Molecular Formula: C23H27FN4O2 Molecular Weight (g/mol): 410.493 MDL Number: MFCD00274576 InChI Key: RAPZEAPATHNIPO-UHFFFAOYSA-N Synonym: risperidone,risperidal,risperdal,risperdal consta,rispolept,risperin,rispolin,sequinan,risperidona,risperidonum PubChem CID: 5073 ChEBI: CHEBI:8871 IUPAC Name: 3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one SMILES: CC1=C(C(=O)N2CCCCC2=N1)CCN3CCC(CC3)C4=NOC5=C4C=CC(=C5)F
R&D Systems™ Risperidone
CAS: 106266-06-2 Molecular Formula: C23H27FN4O2