accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOONHNHNHOHBr

CAS RN 107756-30-9

IUPAC Name: N-[5-(4,5-dihydro-1H-imidazol-2-yl)-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl]methanesulfonamide hydrobromide
Synonyms: N-[5-(4,5-dihydro-1H-imidazol-2-yl)-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl]methanesulfonamide hydrobromide
Molecular Weight (g/mol): 390.3
Molecular Formula: C14H20BrN3O3S
InChi Key: LRFLWCZMTGTUEP-UHFFFAOYNA-N
SMILES: Br.CS(=O)(=O)NC1=C(O)C=CC2=C1CCCC2C1=NCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results