Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ A 61603 hydrobromide
GREENER_CHOICESDP

Catalog No. 105250 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-525-0 50 mg
10-521-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-525-0 Supplier R&D Systems™ Supplier No. 1052/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. α1A agonist

Potent α-adrenoceptor agonist that is at least 35-fold more potent at α1A than at α1B or α1D sites. Induces dose response increases in spontaneous Ca2+ transients in rat ventricular myocytes in vitro (EC50 = 6.9 nmol/L).

Chemical Identifiers

CAS 107756-30-9
Molecular Formula C14H20BrN3O3S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 390.29
Purity >98%
Molecular Weight (g/mol) 390.296
Chemical Name or Material A 61603 hydrobromide
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.