CAS RN 108-87-2
Methylcyclohexane, 99%, extra pure
CAS: 108-87-2 Molecular Formula: C7H14 Molecular Weight (g/mol): 98.19 MDL Number: MFCD00001497 InChI Key: UAEPNZWRGJTJPN-UHFFFAOYSA-N Synonym: cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan PubChem CID: 7962 IUPAC Name: methylcyclohexane SMILES: CC1CCCCC1
Methylcyclohexane, 99%
CAS: 108-87-2 Molecular Formula: C7H14 Molecular Weight (g/mol): 98.19 MDL Number: MFCD00001497 InChI Key: UAEPNZWRGJTJPN-UHFFFAOYSA-N Synonym: cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan PubChem CID: 7962 IUPAC Name: methylcyclohexane SMILES: CC1CCCCC1
Methylcyclohexane, 99%, for spectroscopy
CAS: 108-87-2 Molecular Formula: C7H14 Molecular Weight (g/mol): 98.19 MDL Number: MFCD00001497 InChI Key: UAEPNZWRGJTJPN-UHFFFAOYSA-N Synonym: cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan PubChem CID: 7962 IUPAC Name: methylcyclohexane SMILES: CC1CCCCC1
Methylcyclohexane, 99%, Spectrophotometric Grade, Solstice
CAS: 108-87-2 Molecular Formula: C7H14 Molecular Weight (g/mol): 98.19 MDL Number: MFCD00001497 InChI Key: UAEPNZWRGJTJPN-UHFFFAOYSA-N Synonym: cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan PubChem CID: 7962 IUPAC Name: methylcyclohexane SMILES: CC1CCCCC1
Methylcyclohexane, 99+%
CAS: 108-87-2 Molecular Formula: C7H14 Molecular Weight (g/mol): 98.19 MDL Number: MFCD00001497 InChI Key: UAEPNZWRGJTJPN-UHFFFAOYSA-N Synonym: cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan PubChem CID: 7962 IUPAC Name: methylcyclohexane SMILES: CC1CCCCC1
Methylcyclohexane, 98+%, Extra Dry, AcroSeal™
CAS: 108-87-2 Molecular Formula: C7H14 Molecular Weight (g/mol): 98.19 MDL Number: MFCD00001497 InChI Key: UAEPNZWRGJTJPN-UHFFFAOYSA-N Synonym: cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan PubChem CID: 7962 IUPAC Name: methylcyclohexane SMILES: CC1CCCCC1
Methylcyclohexane, ≥99.5% (GC), MilliporeSigma™ Supelco™
Methylcyclohexane [MCH] is a clear organic flammable liquid, mostly used as a solvent. During the development of chemical kinetic mechanisms for other cycloalkanes, MCH is considered as reaction starting point.
Methylcyclohexane, 99%, Reagent Grade, Solstice
CAS: 108-87-2 Molecular Formula: C7H14 Molecular Weight (g/mol): 98.19 MDL Number: MFCD00001497 InChI Key: UAEPNZWRGJTJPN-UHFFFAOYSA-N Synonym: cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan PubChem CID: 7962 IUPAC Name: methylcyclohexane SMILES: CC1CCCCC1
Methylcyclohexane, ≥99.5% (GC), MilliporeSigma™ Supelco™
Methylcyclohexane [MCH] is a clear organic flammable liquid, mostly used as a solvent. During the development of chemical kinetic mechanisms for other cycloalkanes, MCH is considered as reaction starting point.
Methylcyclohexane 99.0+%, TCI America™
CAS: 108-87-2 Molecular Formula: C7H14 Molecular Weight (g/mol): 98.19 MDL Number: MFCD00001497 InChI Key: UAEPNZWRGJTJPN-UHFFFAOYSA-N Synonym: cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan PubChem CID: 7962 IUPAC Name: methylcyclohexane SMILES: CC1CCCCC1
Methylcyclohexane 99.0+%, TCI America™
CAS: 108-87-2 Molecular Formula: C7H14 Molecular Weight (g/mol): 98.19 MDL Number: MFCD00001497 InChI Key: UAEPNZWRGJTJPN-UHFFFAOYSA-N Synonym: cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan PubChem CID: 7962 IUPAC Name: methylcyclohexane SMILES: CC1CCCCC1
Methylcyclohexane 99.0+%, TCI America™
CAS: 108-87-2 Molecular Formula: C7H14 Molecular Weight (g/mol): 98.19 MDL Number: MFCD00001497 InChI Key: UAEPNZWRGJTJPN-UHFFFAOYSA-N Synonym: cyclohexane, methyl,hexahydrotoluene,cyclohexylmethane,toluene hexahydride,hexahydroxytoluene,sextone b,toluene, hexahydro,metylocykloheksan,methyl cyclohexane,methylcyclohexan PubChem CID: 7962 IUPAC Name: methylcyclohexane SMILES: CC1CCCCC1
Methylcyclohexane, SPEX CertiPrep™
CAS: 108-87-2 Molecular Formula: C7H14 Molecular Weight (g/mol): 98.19 MDL Number: MFCD00001497 InChI Key: UAEPNZWRGJTJPN-UHFFFAOYSA-N PubChem CID: 7962 IUPAC Name: methylcyclohexane SMILES: CC1CCCCC1