Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylcyclohexane, 99%, Spectrophotometric Grade, Solstice

Catalog No. 60048491 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-491 2 L Glass bottle
60-048-490 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-048-491 Supplier Solstice Supplier No. 2596912L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Spectrophotometric Grade, 99%

  • Spectrophotometric grade solvents are high purity solvents made specifically for applications using spectrophotometers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-87-2
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00001497
InChI Key UAEPNZWRGJTJPN-UHFFFAOYSA-N
Synonym cyclohexane, methyl, hexahydrotoluene, cyclohexylmethane, toluene hexahydride, hexahydroxytoluene, sextone b, toluene, hexahydro, metylocykloheksan, methyl cyclohexane, methylcyclohexan
PubChem CID 7962
IUPAC Name methylcyclohexane
SMILES CC1CCCCC1

Specifications

Melting Point -126°C
Density 0.77g/mL at 25°C
Boiling Point 100°C to 102°C
Flash Point −4°C (24.8°F)
Packaging Glass bottle
Refractive Index 1.422
Linear Formula C6H11CH3
Quantity 2 L
UN Number UN2296
Beilstein 505972
Formula Weight 98.19g/mol
Percent Purity 99%
Grade Sprectrophotometric
Chemical Name or Material Methylcyclohexane
Show More Show Less
Hazard Statement P210-P301 + P310-P331-P370 + P378-P403 + P235
Precautionary Statement H225-H304-H315-H336-H410
DOTInformation 3/PGII
EINECSNumber 203-624-3
RTECSNumber GV6125000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.