CAS RN 108179-91-5
IUPAC Name:
8-bromo-3-methyl-5-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepin-7-ol hydrobromide
Synonyms:
8-bromo-3-methyl-5-phenyl-1,2,4,5-tetrahydro-3-benzazepin-7-ol hydrobromide
Molecular Weight (g/mol):
413.15
Molecular Formula:
C17H19Br2NO
InChi Key:
SDQJYYGODYRPBR-UHFFFAOYNA-N
SMILES:
Br.CN1CCC2=CC(Br)=C(O)C=C2C(C1)C1=CC=CC=C1
1
–
1
of
1
results
1
R&D Systems™ SKF 83566 hydrobromide
CAS: 108179-91-5 Molecular Formula: C17H19Br2NO