Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SKF 83566 hydrobromide
GREENER_CHOICESDP

Catalog No. 158610 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-861-0 10 mg
15-865-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 15-861-0 Supplier R&D Systems™ Supplier No. 1586/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, selective D1-like antagonist

Potent and selective D1-like dopamine receptor antagonist (Ki∽ 0.56 nM for D1; KB = 2 μM for D2). Also antagonist at the vascular 5-HT2 receptor (Ki = 11 nM). Displays selective inhibition of adenylyl cyclase 2 (AC2); inactive against AC1 or AC5. Centrally active following systemic administration in vivo.

Chemical Identifiers

CAS 108179-91-5
Molecular Formula C17H19Br2NO

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 413.15
Purity >98%
Molecular Weight (g/mol) 413.153
Chemical Name or Material SKF 83566 hydrobromide
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.