CAS RN 1082739-92-1
IUPAC Name:
3-(2H-1,3-benzodioxol-5-yl)-N-(1-phenylethyl)-[1,2]oxazolo[5,4-c]pyridin-5-amine
Synonyms:
3-(2H-1,3-benzodioxol-5-yl)-N-(1-phenylethyl)-[1,2]oxazolo[5,4-c]pyridin-5-amine
Molecular Weight (g/mol):
359.39
Molecular Formula:
C21H17N3O3
InChi Key:
DAAGOCSHXCDWDY-UHFFFAOYNA-N
SMILES:
CC(NC1=NC=C2ON=C(C2=C1)C1=CC2=C(OCO2)C=C1)C1=CC=CC=C1