CAS RN 108321-42-2
G418 Sulfate (White to Off-white Powder), Fisher BioReagents
CAS: 108321-42-2 Molecular Formula: C20H44N4O18S2 Molecular Weight (g/mol): 692.70 MDL Number: MFCD00058314 InChI Key: UHEPSJJJMTWUCP-KETIWVBHNA-N PubChem CID: 134129582 IUPAC Name: (2R,3S,4R,5R,6S)-5-amino-6-{[(1R,2S,3S,4R,6S)-4,6-diamino-3-{[(2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-2-(1-hydroxyethyl)oxane-3,4-diol; bis(sulfuric acid) SMILES: OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O
Thermo Scientific Chemicals G418 sulfate, for biochemistry
CAS: 108321-42-2 Molecular Formula: C20H44N4O18S2 Molecular Weight (g/mol): 692.70 MDL Number: MFCD00058314 InChI Key: UHEPSJJJMTWUCP-KETIWVBHNA-N PubChem CID: 134129582 SMILES: OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O
G418 disulfate, 50mg/ml solution
CAS: 108321-42-2 Molecular Formula: C20H44N4O18S2 Molecular Weight (g/mol): 692.70 MDL Number: MFCD00058314 InChI Key: UHEPSJJJMTWUCP-KETIWVBHNA-N Synonym: Geneticin PubChem CID: 134129582 IUPAC Name: (2R,3S,4R,5R,6S)-5-amino-6-{[(1R,2S,3S,4R,6S)-4,6-diamino-3-{[(2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-2-(1-hydroxyethyl)oxane-3,4-diol; bis(sulfuric acid) SMILES: OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O
MP Biomedicals™ G418 Disulfate PhytoPure™
G418 Disulfate PhytoPure™ is an aminoglycoside related to gentamicin that acts as an antibiotic and is toxic to bacteria, yeast, higher plants, mammalian cells, protozoans and helminths.
G418 disulfate, Cell Culture Reagent
CAS: 108321-42-2 Molecular Formula: C20H44N4O18S2 Molecular Weight (g/mol): 692.70 MDL Number: MFCD00058314 InChI Key: UHEPSJJJMTWUCP-KETIWVBHNA-N Synonym: Geneticin disulfate PubChem CID: 134129582 SMILES: OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O
G418 Disulfate 90.0+%, TCI America™
CAS: 108321-42-2 Molecular Formula: C20H44N4O18S2 Molecular Weight (g/mol): 692.70 MDL Number: MFCD00058314 InChI Key: UHEPSJJJMTWUCP-KETIWVBHNA-N Synonym: g418 disulfate PubChem CID: 134129582 IUPAC Name: (2R,3S,4R,5R,6S)-5-amino-6-{[(1R,2S,3S,4R,6S)-4,6-diamino-3-{[(2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-2-(1-hydroxyethyl)oxane-3,4-diol; bis(sulfuric acid) SMILES: OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O
MilliporeSigma™ Calbiochem™ G 418 Sulfate, Sterile-Filtered Solution in PBS, Cell Culture-Tested
The optimal concentration of G418 for selection of resistance will vary according to the organism and/or cell type under investigation
R&D Systems™ G418 disulfate salt
CAS: 108321-42-2 Molecular Formula: C20H44N4O18S2