Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ G418 disulfate salt
SDP

Catalog No. 4131100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-311-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 41-311-00 Supplier R&D Systems™ Supplier No. 4131/100

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Aminoglycoside antibiotic; used in cell culture

G418 disulfate salt is an aminoglycosidic antibiotic. Inhibits protein synthesis in both prokaryotic and eukaryotic cells. Used for selection of eukaryotic cells transfected with the neo gene.

This product has been cell culture tested.

Chemical Identifiers

CAS 108321-42-2
Molecular Formula C20H44N4O18S2

Specifications

Quantity 100 mg
Product Line Tocris
Formula Weight 692.71
Product Type G418 Disulfate
Purity >98%
Molecular Weight (g/mol) 692.70
Chemical Name or Material G418 disulfate salt
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.