CAS RN 110044-82-1
IUPAC Name:
(2S)-2-[(2S)-2-acetamido-4-methylpentanamido]-4-methyl-N-[(2S)-1-oxohexan-2-yl]pentanamide
Synonyms:
(2S)-2-[(2S)-2-acetamido-4-methylpentanamido]-4-methyl-N-[(2S)-1-oxohexan-2-yl]pentanamide
Molecular Weight (g/mol):
383.53
Molecular Formula:
C20H37N3O4
InChi Key:
FMYKJLXRRQTBOR-BZSNNMDCSA-N
SMILES:
CCCC[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(C)C)NC(C)=O)C=O
1
–
3
of
3
results
1
MilliporeSigma™ Calbiochem™ ALLN
ALLN, CAS 110044-82-1, is a cell-permeable inhibitor of calpain I (Ki = 190 nM), calpain II (Ki = 220 nM), cathepsin B (Ki = 150 nM), and cathepsin L (Ki = 500 pM).
2
MilliporeSigma™ Calbiochem™ InSolution™ ALLN
A 10mm (5mg/1.30 ml) solution of ALLN (Cat. No. 208719) in DMSO.
3
Thermo Scientific Chemicals Calpain Inhibitor I, 95+%
CAS: 110044-82-1 Molecular Formula: C20H37N3O4 Synonym: Ac-LLnL-CHO; ALLN