CAS RN 111-58-0
IUPAC Name:
(9Z)-N-(2-hydroxyethyl)octadec-9-enamide
Synonyms:
oleoylethanolamide oleamide mea N-(hydroxyethyl) oleamide N-oleoyl ethanolamide
Molecular Weight (g/mol):
325.54
Molecular Formula:
C20H39NO2
InChi Key:
BOWVQLFMWHZBEF-KTKRTIGZSA-N
SMILES:
CCCCCCCC\C=C/CCCCCCCC(=O)NCCO
1
–
1
of
1
results
1
Tocris Bioscience™ Oleylethanolamide
CAS: 111-58-0 Molecular Formula: C20H39NO2