CAS RN 111-84-2
n-Nonane, 99%, pure
CAS: 111-84-2 MDL Number: MFCD00009574 InChI Key: BKIMMITUMNQMOS-UHFFFAOYSA-N Synonym: n-nonane,shellsol 140,nonyl hydride,nonan,heptane, ethyl,unii-t9w3vh6g10,ccris 6081,hsdb 107,dsstox_cid_5796,nonane, analytical standard PubChem CID: 8141 ChEBI: CHEBI:32892 IUPAC Name: nonane SMILES: CCCCCCCCC
n-Nonane, 99%
CAS: 111-84-2 Molecular Formula: C9H20 Molecular Weight (g/mol): 128.259 MDL Number: MFCD00009574 InChI Key: BKIMMITUMNQMOS-UHFFFAOYSA-N Synonym: n-nonane,shellsol 140,nonyl hydride,nonan,heptane, ethyl,unii-t9w3vh6g10,ccris 6081,hsdb 107,dsstox_cid_5796,nonane, analytical standard PubChem CID: 8141 ChEBI: CHEBI:32892 IUPAC Name: nonane SMILES: CCCCCCCCC
n-Nonane, 99%, anhydrous, AcroSeal™
CAS: 111-84-2 Molecular Formula: C9H20 Molecular Weight (g/mol): 128.26 InChI Key: BKIMMITUMNQMOS-UHFFFAOYSA-N Synonym: n-nonane,shellsol 140,nonyl hydride,nonan,heptane, ethyl,unii-t9w3vh6g10,ccris 6081,hsdb 107,dsstox_cid_5796,nonane, analytical standard PubChem CID: 8141 ChEBI: CHEBI:32892 IUPAC Name: nonane SMILES: CCCCCCCCC
Nonane, 99%, Reagent Grade, Solstice
CAS: 111-84-2 Molecular Formula: C9H20 Molecular Weight (g/mol): 128.259 MDL Number: MFCD00009574 InChI Key: BKIMMITUMNQMOS-UHFFFAOYSA-N Synonym: n-nonane,shellsol 140,nonyl hydride,nonan,heptane, ethyl,unii-t9w3vh6g10,ccris 6081,hsdb 107,dsstox_cid_5796,nonane, analytical standard PubChem CID: 8141 ChEBI: CHEBI:32892 IUPAC Name: nonane SMILES: CCCCCCCCC
Nonane, Analytical Standard, MilliporeSigma™ Supelco™
For Gas Chromatography (GC) Analysis
Nonane 99.5+%, TCI America™
CAS: 111-84-2 Molecular Formula: C9H20 Molecular Weight (g/mol): 128.259 MDL Number: MFCD00009574 InChI Key: BKIMMITUMNQMOS-UHFFFAOYSA-N Synonym: n-nonane,shellsol 140,nonyl hydride,nonan,heptane, ethyl,unii-t9w3vh6g10,ccris 6081,hsdb 107,dsstox_cid_5796,nonane, analytical standard PubChem CID: 8141 ChEBI: CHEBI:32892 IUPAC Name: nonane SMILES: CCCCCCCCC
Nonane 98.0+%, TCI America™
CAS: 111-84-2 Molecular Formula: C9H20 Molecular Weight (g/mol): 128.259 MDL Number: MFCD00009574 InChI Key: BKIMMITUMNQMOS-UHFFFAOYSA-N Synonym: n-nonane,shellsol 140,nonyl hydride,nonan,heptane, ethyl,unii-t9w3vh6g10,ccris 6081,hsdb 107,dsstox_cid_5796,nonane, analytical standard PubChem CID: 8141 ChEBI: CHEBI:32892 IUPAC Name: nonane SMILES: CCCCCCCCC
n-Nonane 99.7%, Thermo Scientific™
CAS: 111-84-2 Molecular Formula: C9H20 Molecular Weight (g/mol): 128.259 InChI Key: BKIMMITUMNQMOS-UHFFFAOYSA-N Synonym: n-nonane,shellsol 140,nonyl hydride,nonan,heptane, ethyl,unii-t9w3vh6g10,ccris 6081,hsdb 107,dsstox_cid_5796,nonane, analytical standard PubChem CID: 8141 ChEBI: CHEBI:32892 IUPAC Name: nonane SMILES: CCCCCCCCC
n-Nonane, SPEX CertiPrep™
CAS: 111-84-2 Molecular Formula: C9H20 Molecular Weight (g/mol): 128.259 InChI Key: BKIMMITUMNQMOS-UHFFFAOYSA-N PubChem CID: 8141 ChEBI: CHEBI:32892 IUPAC Name: nonane SMILES: CCCCCCCCC