Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Nonane 99.7%, Thermo Scientific™

Catalog No. AC264310010
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC264310010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC264310010 Supplier Thermo Scientific Chemicals Supplier No. 264310010

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-84-2
Molecular Formula C9H20
Molecular Weight (g/mol) 128.259
InChI Key BKIMMITUMNQMOS-UHFFFAOYSA-N
Synonym n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard
PubChem CID 8141
ChEBI CHEBI:32892
IUPAC Name nonane
SMILES CCCCCCCCC

Specifications

CAS Min % 99.6
CAS Max % 100.0
Color Colorless
Density 0.71
Infrared Spectrum Authentic
Refractive Index 1.4040 to 1.4060 (20°C, 589nm)
Linear Formula CH3(CH2)7CH3
Quantity 1 L
Beilstein 01, 165
Formula Weight 128.26
Percent Purity ≥99.6%
Physical Form Liquid
Chemical Name or Material n-Nonane, 99.7%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.