accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 111662-64-7

OOOOOOOHONH(S)CH3CH3H3CCH3CH3H3C

CAS RN 111662-64-7

IUPAC Name: (2S)-5-(tert-butoxy)-4-[(tert-butoxy)carbonyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoic acid
Synonyms: (2S)-5-(tert-butoxy)-4-(tert-butoxycarbonyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-oxopentanoic acid
Molecular Weight (g/mol): 525.6
Molecular Formula: C29H35NO8
InChi Key: XJRUHYXXHXECDI-QHCPKHFHSA-N
SMILES: CC(C)(C)OC(=O)C(C[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O)C(=O)OC(C)(C)C
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)