CAS RN 1118807-13-8
IUPAC Name:
N-(6-chloro-1,3-benzothiazol-2-yl)-3-(3,4-dimethoxyphenyl)propanamide
Synonyms:
N-(6-chloro-1,3-benzothiazol-2-yl)-3-(3,4-dimethoxyphenyl)propanamide
Molecular Weight (g/mol):
376.86
Molecular Formula:
C18H17ClN2O3S
InChi Key:
LXFKEVQQSKQXPR-UHFFFAOYSA-N
SMILES:
COC1=C(OC)C=C(CCC(=O)NC2=NC3=CC=C(Cl)C=C3S2)C=C1
1
–
1
of
1
results
1
Tocris Bioscience™ KY 02111
Inhibits canonical Wnt signaling. Promotes differentiation of human ESCs and iPSCs into cardiomyocytes