accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)(S)HO(R)(R)CH3(R)OH(S)HOH2C(E)(E)(Z)

CAS RN 112965-21-6

IUPAC Name: (1R,3S,5Z)-5-{2-[(1R,4E,7aR)-1-[(2R,3E,5S)-5-cyclopropyl-5-hydroxypent-3-en-2-yl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexane-1,3-diol
Synonyms: calcipotriol
Molecular Weight (g/mol): 412.61
Molecular Formula: C27H40O3
InChi Key: LWQQLNNNIPYSNX-OUSHRDKSSA-N
SMILES: C[C@H](\C=C\[C@@H](O)C1CC1)[C@H]1CCC2\C(CCC[C@]12C)=C\C=C1\C[C@@H](O)C[C@H](O)C1=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)