Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Calcipotriol
GREENER_CHOICESDP

Catalog No. 270050 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-005-0 50 mg
27-001-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-005-0 Supplier R&D Systems™ Supplier No. 2700/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Vitamin D3 analog

Calcipotriol is a vitamin D3 analog that displays minimal effects on calcium homeostasis. Regulates cell differentiation and proliferation; exhibits antiproliferative activity against human HL-60, HL60/MX2, keratinocytes, MCF-7, T47D, SCC-25, and mouse WEHI-3 cancer cell lines. Calcipotriol suppresses skin cancer development in mice in a thymic stromal lymphopoietin (TSLP)-dependent manner. Calcipotriol promotes the differentiation of keratinocytes. In human T cells and in psoriatic skin, Calcipotriol increases IL-10 secretion, induces secretion of type 2 cytokines by T cells and modulates the differentiation of dendritic cells and T cells.

Chemical Identifiers

CAS 112965-21-6
Molecular Formula C27H40O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 412.6
Purity >98%
Molecular Weight (g/mol) 412.61
Chemical Name or Material Calcipotriol
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.