CAS RN 113170-86-8
IUPAC Name:
2-hydroxyethane-1-sulfonic acid; 6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine
Synonyms:
isethionic acid; lamotrigine
Molecular Weight (g/mol):
382.22
Molecular Formula:
C11H13Cl2N5O4S
InChi Key:
CJIDZLNMKONKAD-UHFFFAOYSA-N
SMILES:
OCCS(O)(=O)=O.NC1=NC(N)=C(N=N1)C1=C(Cl)C(Cl)=CC=C1
1
–
1
of
1
results
1
R&D Systems™ Lamotrigine isethionate
CAS: 113170-86-8 Molecular Formula: C11H13Cl2N5O4S