accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 113305-56-9

OHNOOI

CAS RN 113305-56-9

IUPAC Name: 4-iodo-3-nitrophenol
Synonyms: 4-iodo-3-nitrophenol
Molecular Weight (g/mol): 265.01
Molecular Formula: C6H4INO3
InChi Key: QCKXOEOOXJUACP-UHFFFAOYSA-N
SMILES: OC1=CC(=C(I)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results