CAS RN 1135278-54-4
IUPAC Name:
(1-{[(7-bromo-2,3-dioxo-1,2,3,4-tetrahydroquinoxalin-5-yl)methyl]amino}ethyl)phosphonic acid hydrochloride
Synonyms:
1-{[(7-bromo-2,3-dioxo-1,4-dihydroquinoxalin-5-yl)methyl]amino}ethylphosphonic acid hydrochloride
Molecular Weight (g/mol):
414.58
Molecular Formula:
C11H14BrClN3O5P
InChi Key:
MZQQZBPMRPDKTB-UHFFFAOYNA-N
SMILES:
Cl.CC(NCC1=C2NC(=O)C(=O)NC2=CC(Br)=C1)P(O)(O)=O
1
–
1
of
1
results
1
R&D Systems™ CGP 78608 hydrochloride
CAS: 1135278-54-4 Molecular Formula: C11H14BrClN3O5P