accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1135280-28-2

IUPAC Name: N-[2-({[3-(4-chlorophenyl)prop-2-en-1-yl](methyl)amino}methyl)phenyl]-4-methoxybenzene-1-sulfonamide; phosphoric acid
Synonyms: N-[2-({[3-(4-chlorophenyl)prop-2-en-1-yl](methyl)amino}methyl)phenyl]-4-methoxybenzenesulfonamide; phosphoric acid
Molecular Weight (g/mol): 554.98
Molecular Formula: C24H28ClN2O7PS
InChi Key: XRQHWVVDNMJDEQ-UHFFFAOYSA-N
SMILES: OP(O)(O)=O.COC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=C1CN(C)CC=CC1=CC=C(Cl)C=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results