CAS RN 114-49-8
IUPAC Name:
(1S,2R,4S,5S)-9-methyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-7-yl (2S)-3-hydroxy-2-phenylpropanoate hydrobromidyl
Synonyms:
(1S,2R,4S,5S)-9-methyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-7-yl (2S)-3-hydroxy-2-phenylpropanoate hydrobromidyl
Molecular Weight (g/mol):
383.26
Molecular Formula:
C17H21BrNO4
InChi Key:
XUZPYCBMTSSDHK-AKTDCHNFSA-N
SMILES:
[Br].CN1[C@H]2CC(C[C@H]1[C@@H]1O[C@H]21)OC(=O)[C@H](CO)C1=CC=CC=C1
1
–
1
of
1
results
1
Tocris Bioscience™ Scopolamine hydrobromide
CAS: 114-49-8 Molecular Formula: C17H22BrNO4