Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Scopolamine hydrobromide
GREENER_CHOICESDP

Catalog No. 14141GR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-141-GR 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-141-GR Supplier Tocris Bioscience™ Supplier No. 1414/1G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Non-selective muscarinic antagonist

Non-selective muscarinic antagonist. Widely used clinically to treat motion sickness.

Chemical Identifiers

CAS 114-49-8
Molecular Formula C17H22BrNO4

Specifications

Quantity 1 g
Formula Weight 384.27
Physical Form Solid
Purity >99%
Molecular Weight (g/mol) 384.27
Chemical Name or Material Scopolamine hydrobromide
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.