CAS RN 1160514-60-2
IUPAC Name:
ethyl 1-[4-(2,3,3-trichloroprop-2-enamido)phenyl]-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate
Synonyms:
ethyl 1-[4-(2,3,3-trichloroprop-2-enamido)phenyl]-5-(trifluoromethyl)pyrazole-4-carboxylate
Molecular Weight (g/mol):
456.63
Molecular Formula:
C16H11Cl3F3N3O3
InChi Key:
RZHGONNSASQOAY-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C(N(N=C1)C1=CC=C(NC(=O)C(Cl)=C(Cl)Cl)C=C1)C(F)(F)F
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ TRPC3 Channel Inhibitor, Pyr3
The TRPC3 Channel Inhibitor, Pyr3, also referenced under CAS 1160514-60-2, controls the biological activity of TRPC3 Channel. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
2
Tocris Bioscience™ Pyr3
CAS: 1160514-60-2 Molecular Formula: C16H11Cl3F3N3O3