accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHOOOHOHOH

CAS RN 117-39-5

IUPAC Name: 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one
Synonyms: quercetin 槲皮素
Molecular Weight (g/mol): 302.24
Molecular Formula: C15H10O7
InChi Key: REFJWTPEDVJJIY-UHFFFAOYSA-N
SMILES: OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC=C(O)C(O)=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)