CAS RN 1173023-85-2
IUPAC Name:
N-{3-chloro-4-[(3-fluorophenyl)methoxy]phenyl}-6-(2-{[(2-methanesulfonylethyl)amino]methyl}-1,3-thiazol-4-yl)quinazolin-4-amine dihydrochloride
Synonyms:
N-{3-chloro-4-[(3-fluorophenyl)methoxy]phenyl}-6-(2-{[(2-methanesulfonylethyl)amino]methyl}-1,3-thiazol-4-yl)quinazolin-4-amine dihydrochloride
Molecular Weight (g/mol):
671.02
Molecular Formula:
C28H27Cl3FN5O3S2
InChi Key:
WIMITXDBYLKRKB-UHFFFAOYSA-N
SMILES:
Cl.Cl.CS(=O)(=O)CCNCC1=NC(=CS1)C1=CC=C2N=CN=C(NC3=CC(Cl)=C(OCC4=CC(F)=CC=C4)C=C3)C2=C1
1
–
1
of
1
results
1
R&D Systems™ GW 583340 dihydrochloride
CAS: 1173023-85-2 Molecular Formula: C28H27Cl3FN5O3S2