Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ GW 583340 dihydrochloride
GREENER_CHOICESDP

Catalog No. 223950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-395-0 50 mg
22-391-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-395-0 Supplier R&D Systems™ Supplier No. 2239/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent dual EGFR/ErbB2 inhibitor; orally active

Potent dual EGFR/ErbB2 tyrosine kinase inhibitor (IC50 values are 0.01 and 0.014 μM respectively). Selectively inhibits growth of human tumor cells overexpressing EGFR and ErbB2 (IC50 values are 0.11 μM for inhibition of HN5, N87 and BT474 tumor cell lines vs. > 30 μM for inhibition of non-tumor cell line HFF). Inhibits tumor growth in vivo by ∽ 80% in a murine xenograft model following oral administration.

Chemical Identifiers

CAS 1173023-85-2
Molecular Formula C28H27Cl3FN5O3S2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 671.03
Purity >97%
Molecular Weight (g/mol) 671.024
Chemical Name or Material GW 583340 dihydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.