accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONONHNOF

CAS RN 1174043-16-3

IUPAC Name: 4-{[4-fluoro-3-(4-methoxypiperidine-1-carbonyl)phenyl]methyl}-1,2-dihydrophthalazin-1-one
Synonyms: 4-{[4-fluoro-3-(4-methoxypiperidine-1-carbonyl)phenyl]methyl}-2H-phthalazin-1-one
Molecular Weight (g/mol): 395.43
Molecular Formula: C22H22FN3O3
InChi Key: HYNBNUYQTQIHJK-UHFFFAOYSA-N
SMILES: COC1CCN(CC1)C(=O)C1=C(F)C=CC(CC2=NNC(=O)C3=CC=CC=C23)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)