CAS RN 117570-53-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Tocris Bioscience™ DMXAA
Tocris Bioscience™ DMXAA
mSTING agonist; induces antitumor immunological responses