accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 117690-79-6

IUPAC Name: 1-(5-ethyl-2-hydroxy-4-{[6-methyl-6-(2H-1,2,3,4-tetrazol-5-yl)heptyl]oxy}phenyl)ethan-1-one
Synonyms: 1-(5-ethyl-2-hydroxy-4-{[6-methyl-6-(2H-1,2,3,4-tetrazol-5-yl)heptyl]oxy}phenyl)ethanone
Molecular Weight (g/mol): 360.46
Molecular Formula: C19H28N4O3
InChi Key: WCGXJPFHTHQNJL-UHFFFAOYSA-N
SMILES: CCC1=CC(C(C)=O)=C(O)C=C1OCCCCCC(C)(C)C1=NNN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results