accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 117857-93-9

IUPAC Name: 2-[amino(carboxy)methyl]cyclopropane-1-carboxylic acid
Synonyms: 2-[amino(carboxy)methyl]cyclopropane-1-carboxylic acid
Molecular Weight (g/mol): 159.14
Molecular Formula: C6H9NO4
InChi Key: GZOVEPYOCJWRFC-UHFFFAOYNA-N
SMILES: NC(C1CC1C(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results