CAS RN 1181083-81-7
CAS RN 1181083-81-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ CLP 290
K+/Cl- cotransporter 2 (KCC2) activator; orally bioavailable prodrug of CLP 257 (Cat. No.5242)