accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 118290-26-9

IUPAC Name: N-(2-{2-[(dipropylamino)methyl]piperidin-1-yl}ethyl)-10-oxo-2,4,9-triazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3(8),4,6,11,13-hexaene-2-carboxamide
Synonyms: N-(2-{2-[(dipropylamino)methyl]piperidin-1-yl}ethyl)-10-oxo-2,4,9-triazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3(8),4,6,11,13-hexaene-2-carboxamide
Molecular Weight (g/mol): 478.64
Molecular Formula: C27H38N6O2
InChi Key: MZDYABXXPZNUCT-UHFFFAOYNA-N
SMILES: CCCN(CCC)CC1CCCCN1CCNC(=O)N1C2=CC=CC=C2C(=O)NC2=C1N=CC=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results