Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AF-DX 384
GREENER_CHOICESDP

Catalog No. 134550 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-455-0 50 mg
13-451-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 13-455-0 Supplier R&D Systems™ Supplier No. 1345/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent M2/M4 antagonist

Potent M2/M4 selective antagonist (pKi values are 8.22, 8.00, 7.51, 7.18 and 6.27 at human M2, M4, M1, M3 and M5 receptors respectively).

Chemical Identifiers

CAS 118290-26-9
Molecular Formula C27H38N6O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 478.64
Purity >98%
Molecular Weight (g/mol) 478.641
Chemical Name or Material AF-DX 384
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.