CAS RN 118876-58-7
IUPAC Name:
6-nitro-2,3-dioxo-1H,2H,3H,4H-benzo[f]quinoxaline-7-sulfonamide
Synonyms:
NBQX
Molecular Weight (g/mol):
336.28
Molecular Formula:
C12H8N4O6S
InChi Key:
UQNAFPHGVPVTAL-UHFFFAOYSA-N
SMILES:
NS(=O)(=O)C1=C2C(=CC=C1)C1=C(NC(=O)C(=O)N1)C=C2[N+]([O-])=O