CAS RN 1195768-06-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
R&D Systems™ Dabrafenib mesylate
A Tocris product, part of R&D Systems. Potent and selective B-Raf, CDK16 and NEK9 inhibitor; anticancer; also targets human proteins in the SARS-CoV-2 interactome