CAS RN 1203680-76-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ AS 1949490
SH2 domain-containing inositol 5'-phosphatase 2 (SHIP2) inhibitor