accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNOHOHHONH2HCl

CAS RN 120964-45-6

IUPAC Name: 5-{4-amino-1H-imidazo[4,5-c]pyridin-1-yl}-3-(hydroxymethyl)cyclopent-3-ene-1,2-diol hydrochloride
Synonyms: 5-{4-aminoimidazo[4,5-c]pyridin-1-yl}-3-(hydroxymethyl)cyclopent-3-ene-1,2-diol hydrochloride
Molecular Weight (g/mol): 298.73
Molecular Formula: C12H15ClN4O3
InChi Key: UNSKMHKAFPRFTI-UHFFFAOYNA-N
SMILES: Cl.NC1=C2N=CN(C3C=C(CO)C(O)C3O)C2=CC=N1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results