accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 121034-89-7

IUPAC Name: (5-{[(adamantan-1-yl)methyl]amino}pentyl)trimethylazanium hydrobromide bromide
Synonyms: {5-[(adamantan-1-ylmethyl)amino]pentyl}trimethylazanium hydrobromide bromide
Molecular Weight (g/mol): 454.34
Molecular Formula: C19H38Br2N2
InChi Key: CQTDZUSQSTUZDA-UHFFFAOYSA-M
SMILES: Br.[Br-].C[N+](C)(C)CCCCCNCC12CC3CC(CC(C3)C1)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)