Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ IEM 1460
GREENER_CHOICESDP

Catalog No. 163650 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-365-0 50 mg
16-361-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-365-0 Supplier R&D Systems™ Supplier No. 1636/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Open-channel blocker of AMPA currents; selective for non-GluA2-containing receptors

Voltage-dependent open-channel blocker of AMPA receptors. Selective for GluR2 subunit-lacking (Ca2+-permeable) receptors over GluA2-containing receptors (IC50 values are 2.6 and 1102 μM, respectively). Selectively blocks fast spiking interneuron but not medium spiny projection neuron in mouse striatum. Also blocks NMDA receptor-mediated currents. Anticonvulsant in vivo.

Chemical Identifiers

CAS 121034-89-7
Molecular Formula C19H38Br2N2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 454.33
Purity >98%
Molecular Weight (g/mol) 454.335
Chemical Name or Material IEM 1460
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.