CAS RN 121524-09-2
IUPAC Name:
hydrogen ethyl 2-{[(7S)-7-{[(2R)-2-(3-chlorophenyl)-2-hydroxyethyl]amino}-5,6,7,8-tetrahydronaphthalen-2-yl]oxy}acetate chloride
Synonyms:
amibegron
Molecular Weight (g/mol):
440.36
Molecular Formula:
C22H27Cl2NO4
InChi Key:
NQIZCDQCNYCVAS-RQBPZYBGSA-N
SMILES:
[H+].[Cl-].CCOC(=O)COC1=CC=C2CC[C@@H](CC2=C1)NC[C@H](O)C1=CC=CC(Cl)=C1
1
–
1
of
1
results
1
R&D Systems™ SR 58611A hydrochloride
CAS: 121524-09-2 Molecular Formula: C22H27Cl2NO4